Assay Details
Target Gene Details
Entrez Gene ID: | 84687 |
Gene Name: | protein phosphatase 1 regulatory subunit 9B |
Gene Aliases: |
PPP1R6, PPP1R9, SPINO, Spn |
Location: |
Chr.17:50133735-50151027 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP1R9B | NM_032595.4 | NP_115984.3 | ||
AJ401189.1 | CAC37685.1 | |||
AK054907.1 | ||||
AL713642.1 | CAD28455.2 | |||
BC016162.1 | AAH16162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952339 | Chr.17:50115237 - 50222139 on Build GRCh38 | Deletion | SGCA PPP1R9B HILS1 SAMD14 COL1A1 LOC101927230 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |