Assay Details
Target Gene Details
Entrez Gene ID: | 100130950 |
Gene Name: | uncharacterized LOC100130950 |
Gene Aliases: |
- |
Location: |
Chr.17:5192084-5235636 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100130950 | NR_034082.1 |
Target Gene Details
Entrez Gene ID: | 388325 |
Gene Name: | SLP adaptor and CSK interacting membrane protein |
Gene Aliases: |
C17orf87, UNQ5783 |
Location: |
Chr.17:5208920-5234860 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SCIMP | NM_001271842.1 | NP_001258771.1 | ||
NM_001319190.1 | NP_001306119.1 | |||
NM_207103.3 | NP_996986.1 | |||
XM_017024615.1 | XP_016880104.1 | |||
AK057142.1 | ||||
AK297035.1 | ||||
AK310145.1 | ||||
AY358809.1 | AAQ89169.1 | |||
BC127925.1 | ||||
BC130554.1 | ||||
BC130556.1 | ||||
BC144395.1 | ||||
DQ778084.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain | RPAIN SLC25A11 ARRB2 GP1BA LOC107984973 LOC101928000 LOC100130950 ALOX15 PELP1 LOC101559451 RABEP1 CHRNE PFN1 ZNF232 SPAG7 SLC52A1 INCA1 MED11 VMO1 KIF1C CXCL16 PLD2 ZFP3 SCIMP ZNF594 SMTNL2 CAMTA2 DHX33 C17orf107 LOC102724009 ENO3 PSMB6 USP6 MINK1 LOC101927979 LOC400568 NUP88 MIR6864 ZMYND15 MIR6865 C1QBP RNF167 TM4SF5 GLTPD2 |
dgv3089n100 | Chr.17:5187013 - 5327634 on Build GRCh38 | Gain | LOC100130950 SCIMP RABEP1 ZNF594 |
nsv952114 | Chr.17:5228306 - 5237005 on Build GRCh38 | Deletion | LOC100130950 SCIMP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |