Assay Details
Target Gene Details
Entrez Gene ID: | 642 |
Gene Name: | bleomycin hydrolase |
Gene Aliases: |
BH, BMH |
Location: |
Chr.17:30248195-30292166 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BLMH | NM_000386.3 | NP_000377.1 | ||
AF091082.1 | AAC72951.1 | |||
AK223403.1 | BAD97123.1 | |||
AK301948.1 | ||||
AK312896.1 | ||||
BC003616.2 | AAH03616.1 | |||
BT007018.1 | AAP35664.1 | |||
X92106.1 | CAA63078.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2023 | Chr.17:30245788 - 30260021 on Build GRCh38 | Insertion | BLMH |
esv3582599 | Chr.17:30248965 - 30255400 on Build GRCh38 | Loss | BLMH |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |