Assay Details
Target Gene Details
Entrez Gene ID: | 84316 |
Gene Name: | N(alpha)-acetyltransferase 38, NatC auxiliary subunit |
Gene Aliases: |
LSMD1, PFAAP2 |
Location: |
Chr.17:7856685-7885465 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAA38 | XM_011524037.2 | XP_011522339.1 | ||
XM_017025225.1 | XP_016880714.1 | |||
XM_017025226.1 | XP_016880715.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952120 | Chr.17:7867583 - 7883482 on Build GRCh38 | Duplication | NAA38 |
dgv510n67 | Chr.17:7852429 - 7911448 on Build GRCh38 | Gain | CYB5D1 KDM6B SCARNA21 CHD3 TMEM88 NAA38 |
nsv833351 | Chr.17:7755691 - 7893863 on Build GRCh38 | Loss | CYB5D1 DNAH2 KDM6B CHD3 TMEM88 NAA38 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |