Assay Details
Target Gene Details
Entrez Gene ID: | 11216 |
Gene Name: | A-kinase anchoring protein 10 |
Gene Aliases: |
AKAP-10, D-AKAP-2, D-AKAP2, PRKA10 |
Location: |
Chr.17:19904452-19977856 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AKAP10 | NM_007202.3 | NP_009133.2 | ||
XM_006721432.3 | XP_006721495.1 | |||
AF037439.1 | AAB92260.1 | |||
AK308297.1 | ||||
AK312438.1 | ||||
BC017055.1 | AAH17055.1 | |||
DQ270159.1 | ABB76684.1 | |||
KU178546.1 | ||||
KU178547.1 | ||||
KU178548.1 | ||||
KU178549.1 | ||||
KU178550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1061317 | Chr.17:19047542 - 20322438 on Build GRCh38 | Gain | GRAPL B9D1 RNF112 EPN2-AS1 SNORD3B-1 SNORD3B-2 LOC388436 CCDC144CP ULK2 RPS2P46 LOC102724624 SNORD3C EPN2-IT1 GRAP MAPK7 ALDH3A2 SLC47A1 EPN2 AKAP10 ALDH3A1 SLC47A2 SPECC1 MFAP4 LOC100131943 SNORD3D SNORD3A SNORA59B MIR1180 LOC79999 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |