Assay Details
Target Gene Details
Entrez Gene ID: | 100505591 |
Gene Name: | leucine rich repeat containing 3C |
Gene Aliases: |
- |
Location: |
Chr.17:39927722-39944999 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRC3C | XM_011524158.2 | XP_011522460.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3573064 | Chr.17:39896348 - 40029806 on Build GRCh38 | Gain | MED24 ORMDL3 CSF3 GSDMB SNORD124 PSMD3 LRRC3C MIR6884 GSDMA |