Assay Details
Target Gene Details
Entrez Gene ID: | 3980 |
Gene Name: | DNA ligase 3 |
Gene Aliases: |
LIG2 |
Location: |
Chr.17:34980498-35009169 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIG3 | NM_002311.4 | NP_002302.2 | ||
NM_013975.3 | NP_039269.2 | |||
XM_005257970.3 | XP_005258027.1 | |||
XM_006721896.3 | XP_006721959.1 | |||
XM_017024624.1 | XP_016880113.1 | |||
AK300363.1 | ||||
AK308407.1 | ||||
BC009026.1 | AAH09026.1 | |||
BC068005.1 | AAH68005.1 | |||
BM759573.1 | ||||
DB100562.1 | ||||
U40671.1 | AAA85022.1 | |||
X84740.1 | CAA59230.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)