Assay Details
Target Gene Details
Entrez Gene ID: | 1984 |
Gene Name: | eukaryotic translation initiation factor 5A |
Gene Aliases: |
EIF-5A, EIF5A1, eIF5AI |
Location: |
Chr.17:7306999-7312463 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF5A | NM_001143760.1 | NP_001137232.1 | ||
XM_017024301.1 | XP_016879790.1 | |||
AY129319.1 | AAN17514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion | TMEM95 POLR2A EIF5A MIR195 GPS2 SHBG TMEM256-PLSCR3 TNK1 SENP3-EIF4A1 CTDNEP1 SLC16A11 ACADVL RNASEK-C17orf49 FXR2 SNORA67 TNFSF12 KCTD11 PHF23 C17orf74 BCL6B SLC16A13 DLG4 SPEM1 SENP3 CLDN7 LOC100996842 DVL2 TEKT1 ELP5 FGF11 RNASEK CLEC10A FBXO39 TMEM102 GABARAP EIF4A1 MPDU1 TNFSF12-TNFSF13 CHRNB1 PLSCR3 SOX15 ATP1B2 ZBTB4 SNORA48 TNFSF13 ALOX12P2 MIR324 SLC2A4 C17orf49 TMEM256 SLC35G6 MIR497 ASGR2 SAT2 LOC107983988 ALOX12-AS1 ALOX12 NLGN2 MIR497HG ASGR1 ACAP1 SNORD10 CD68 NEURL4 YBX2 |
esv3639865 | Chr.17:7292786 - 7322537 on Build GRCh38 | Loss | EIF5A GPS2 NEURL4 YBX2 |
nsv509650 | Chr.17:7299079 - 7338714 on Build GRCh38 | Insertion | EIF5A ACAP1 GPS2 NEURL4 |
nsv952118 | Chr.17:7174582 - 7456981 on Build GRCh38 | Deletion | TMEM95 EIF5A GPS2 CHRNB1 PLSCR3 TMEM256-PLSCR3 TNK1 CTDNEP1 MIR324 SLC2A4 ACADVL TMEM256 KCTD11 PHF23 C17orf74 LOC107983988 DLG4 SPEM1 NLGN2 CLDN7 DVL2 ASGR1 ACAP1 ELP5 FGF11 NEURL4 TMEM102 YBX2 GABARAP |
esv2665200 | Chr.17:7091816 - 7333062 on Build GRCh38 | Deletion | PHF23 EIF5A GPS2 ASGR2 DLG4 CLDN7 CTDNEP1 DVL2 ASGR1 ELP5 MIR324 SLC2A4 NEURL4 ACADVL YBX2 GABARAP |
nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss | TMEM95 POLR2A EIF5A MIR195 GPS2 SHBG TMEM256-PLSCR3 TNK1 SENP3-EIF4A1 CTDNEP1 SLC16A11 WRAP53 ACADVL RNASEK-C17orf49 FXR2 SNORA67 TNFSF12 KCTD11 PHF23 C17orf74 BCL6B SLC16A13 DLG4 SPEM1 SENP3 CLDN7 LOC100996842 DVL2 ELP5 FGF11 RNASEK CLEC10A TMEM102 GABARAP EIF4A1 MPDU1 TP53 TNFSF12-TNFSF13 CHRNB1 PLSCR3 SOX15 ATP1B2 ZBTB4 SNORA48 TNFSF13 MIR324 SLC2A4 C17orf49 TMEM256 SLC35G6 MIR497 ASGR2 SAT2 LOC107983988 ALOX12-AS1 ALOX12 NLGN2 MIR497HG ASGR1 ACAP1 SNORD10 CD68 NEURL4 YBX2 |
nsv1963 | Chr.17:7244585 - 7333855 on Build GRCh38 | Deletion | CTDNEP1 EIF5A GPS2 ELP5 SLC2A4 NEURL4 YBX2 CLDN7 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |