Assay Details
Target Gene Details
Entrez Gene ID: | 105369205 |
Gene Name: | uncharacterized LOC105369205 |
Gene Aliases: |
- |
Location: |
Chr.17:20921016-21001059 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105369205 |
Target Gene Details
Entrez Gene ID: | 339260 |
Gene Name: | uncharacterized LOC339260 |
Gene Aliases: |
- |
Location: |
Chr.17:20938520-20982357 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC339260 | NR_123730.1 | |||
DB066214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1064365 | Chr.17:20898426 - 20987403 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv5502n54 | Chr.17:20899465 - 20969018 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2751619 | Chr.17:20559595 - 21010843 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv960441 | Chr.17:20966695 - 20969465 on Build GRCh38 | Duplication |
![]() ![]() |
nsv1059146 | Chr.17:20849194 - 22546654 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3640224 | Chr.17:20957522 - 20982745 on Build GRCh38 | Loss |
![]() ![]() |
nsv457706 | Chr.17:20911453 - 20969018 on Build GRCh38 | Gain |
![]() ![]() |
nsv2006 | Chr.17:20943450 - 20989417 on Build GRCh38 | Deletion |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)