Assay Details
Target Gene Details
Entrez Gene ID: | 7716 |
Gene Name: | vascular endothelial zinc finger 1 |
Gene Aliases: |
DB1, ZNF161 |
Location: |
Chr.17:57971547-57988254 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VEZF1 | NM_007146.2 | NP_009077.2 | ||
XM_005257643.1 | XP_005257700.1 | |||
XM_005257644.4 | XP_005257701.1 | |||
XM_017025017.1 | XP_016880506.1 | |||
XM_017025018.1 | XP_016880507.1 | |||
AA195154.1 | ||||
BX648862.1 | ||||
D28118.1 | BAA05663.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833497 | Chr.17:57929210 - 58108015 on Build GRCh38 | Loss | VEZF1 LOC101929185 CUEDC1 LOC101927666 DYNLL2 SRSF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |