Assay Details
Target Gene Details
Entrez Gene ID: | 84690 |
Gene Name: | spermatogenesis associated 22 |
Gene Aliases: |
NYD-SP20, NYDSP20 |
Location: |
Chr.17:3440011-3513852 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPATA22 | NM_001170695.1 | NP_001164166.1 | ||
NM_001170696.1 | NP_001164167.1 | |||
NM_001170697.1 | NP_001164168.1 | |||
NM_001170698.1 | NP_001164169.1 | |||
NM_001170699.1 | NP_001164170.1 | |||
NM_001321336.1 | NP_001308265.1 | |||
NM_001321337.1 | NP_001308266.1 | |||
NM_032598.4 | NP_115987.2 | |||
AF367472.1 | AAK53408.1 | |||
AK295299.1 | ||||
AK301892.1 | ||||
AY032684.1 | AAK51120.1 | |||
AY035867.1 | AAK61373.1 | |||
AY035868.1 | AAK61374.1 | |||
BC029483.1 | AAH29483.1 | |||
BG716998.1 | ||||
BI463957.1 | ||||
BP371122.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892950 | Chr.17:3131812 - 3577153 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv574232 | Chr.17:3254715 - 3503170 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2660206 | Chr.17:2679367 - 3519614 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1055740 | Chr.17:3381841 - 3890153 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv521277 | Chr.17:3390044 - 3468885 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv833341 | Chr.17:3352243 - 3521390 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1060071 | Chr.17:3398474 - 3633472 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1059413 | Chr.17:3365789 - 3453922 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)