Assay Details
Target Gene Details
Entrez Gene ID: | 7266 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C7 |
Gene Aliases: |
DJ11, DJC7, TPR2, TTC2 |
Location: |
Chr.17:41976421-42021371 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNAJC7 | NM_001144766.2 | NP_001138238.1 | ||
NM_003315.3 | NP_003306.3 | |||
NR_029431.1 | ||||
XM_011525167.2 | XP_011523469.1 | |||
XM_011525168.2 | XP_011523470.1 | |||
XM_011525169.2 | XP_011523471.1 | |||
XM_017024994.1 | XP_016880483.1 | |||
AB209834.1 | BAD93071.1 | |||
AK291047.1 | ||||
AK296602.1 | ||||
AK298860.1 | ||||
AK316081.1 | ||||
BC003601.1 | AAH03601.1 | |||
BC011837.2 | AAH11837.2 | |||
BC033772.1 | AAH33772.1 | |||
BQ445927.1 | ||||
BX647209.1 | ||||
U46571.1 | AAB36872.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509658 | Chr.17:41927491 - 42002888 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)