Assay Details
Target Gene Details
Entrez Gene ID: | 284021 |
Gene Name: | mast cell immunoglobulin like receptor 1 |
Gene Aliases: |
Allergin-1, C17orf60, MCA-32, MCA32 |
Location: |
Chr.17:64449022-64474009 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MILR1 | XM_017024486.1 | XP_016879975.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |