Assay Details
Target Gene Details
Entrez Gene ID: | 22843 |
Gene Name: | protein phosphatase, Mg2+/Mn2+ dependent 1E |
Gene Aliases: |
CaMKP-N, POPX1, PP2CH, caMKN |
Location: |
Chr.17:58755869-58985617 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPM1E | NM_014906.4 | NP_055721.3 | ||
NR_048561.1 | ||||
AB028995.1 | ||||
AF260269.1 | AAF70325.1 | |||
AF520614.1 | AAM76058.1 | |||
AK289966.1 | ||||
BC136290.1 | ||||
BC151228.1 | ||||
CR749253.1 | CAH18109.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1057312 | Chr.17:58748383 - 58804942 on Build GRCh38 | Gain | PPM1E |
nsv833498 | Chr.17:58652063 - 58815031 on Build GRCh38 | Loss | RAD51C PPM1E TEX14 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |