Assay Details
Target Gene Details
Entrez Gene ID: | 4706 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit AB1 |
Gene Aliases: |
ACP, FASN2A, SDAP |
Location: |
Chr.16:23581012-23596329 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFAB1 | NM_005003.2 | NP_004994.1 | ||
XM_011545856.2 | XP_011544158.2 | |||
AF087660.1 | AAD23566.1 | |||
AK311877.1 | ||||
BC058920.1 | AAH58920.1 | |||
BI603093.1 | ||||
EF036489.1 | ||||
EF036490.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |