Assay Details
Target Gene Details
Entrez Gene ID: | 653319 |
Gene Name: | KIAA0895 like |
Gene Aliases: |
- |
Location: |
Chr.16:67175602-67183980 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KIAA0895L | NM_001040715.1 | NP_001035805.1 | ||
AK094787.1 | ||||
AK096855.1 | ||||
AL834156.1 | CAD38862.1 | |||
BC080183.1 | AAH80183.1 | |||
BX460002.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | HSD11B2 ZDHHC1 LOC100131303 CENPT CTRL C16orf86 PSMB10 KIAA0895L TRADD ATP6V0D1 LOC105369155 SLC12A4 LOC107984813 NOL3 NRN1L TPPP3 NUTF2 PARD6A CARMIL2 ENKD1 PLEKHG4 LRRC29 THAP11 RANBP10 SLC9A5 EDC4 DPEP3 DUS2 LCAT LRRC36 KCTD19 ELMO3 MIR328 FAM65A TSNAXIP1 E2F4 AGRP DDX28 TMEM208 PSKH1 GFOD2 LOC100505942 FBXL8 EXOC3L1 HSF4 CTCF ACD DPEP2 FHOD1 |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | SLC9A5 E2F4 LOC105369155 TMEM208 NOL3 FBXL8 EXOC3L1 HSF4 KIAA0895L B3GNT9 ELMO3 TRADD MIR328 C16orf70 PLEKHG4 LRRC29 FHOD1 |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | E2F4 LOC105369155 TMEM208 NOL3 FBXL8 EXOC3L1 HSF4 KIAA0895L B3GNT9 ELMO3 TRADD MIR328 C16orf70 LRRC29 FHOD1 |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | HSF4 E2F4 LOC105369155 KIAA0895L ELMO3 MIR328 NOL3 FBXL8 EXOC3L1 LRRC29 |
nsv457513 | Chr.16:67181417 - 67282697 on Build GRCh38 | Loss | SLC9A5 E2F4 LOC105369155 KIAA0895L TMEM208 ELMO3 MIR328 EXOC3L1 PLEKHG4 LRRC29 FHOD1 |
nsv524492 | Chr.16:67174030 - 67294550 on Build GRCh38 | Loss | SLC9A5 E2F4 LOC105369155 TMEM208 NOL3 EXOC3L1 KIAA0895L KCTD19 ELMO3 MIR328 PLEKHG4 LRRC29 FHOD1 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | HSD11B2 ZDHHC1 LOC100131303 CENPT CTRL C16orf86 PSMB10 KIAA0895L B3GNT9 TRADD ATP6V0D1 LOC105369155 SLC12A4 LOC107984813 NOL3 NRN1L TPPP3 NUTF2 PARD6A CARMIL2 ENKD1 PLEKHG4 LRRC29 THAP11 RANBP10 SLC9A5 EDC4 DPEP3 DUS2 LCAT LRRC36 KCTD19 ELMO3 MIR328 FAM65A TSNAXIP1 E2F4 AGRP DDX28 TMEM208 PSKH1 GFOD2 LOC100505942 FBXL8 EXOC3L1 HSF4 CTCF ACD C16orf70 DPEP2 FHOD1 |