Assay Details
Target Gene Details
Entrez Gene ID: | 124274 |
Gene Name: | G protein-coupled receptor 139 |
Gene Aliases: |
GPRg1, PGR3 |
Location: |
Chr.16:20031485-20073778 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR139 | NM_001002911.3 | NP_001002911.1 | ||
NM_001318483.1 | NP_001305412.1 | |||
AB196529.1 | BAD97440.1 | |||
AK291384.1 | ||||
AY635179.1 | AAT65818.1 | |||
BC137237.1 | ||||
BC137238.1 | ||||
DN693347.1 | ||||
KR081941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | PDILT ACSM2A CRYM-AS1 TMC7 TMEM159 GDE1 SNX29P1 LOC100190986 CLEC19A LYRM1 KNOP1 COQ7 ZP2 GPR139 ACSM1 LOC102723385 CRYM THUMPD1 IQCK ITPRIPL2 ACSM2B CCP110 ERI2 ANKS4B DNAH3 GPRC5B SYT17 SMG1P3 DCUN1D3 TMC5 NPIPB3 ACSM5 C16orf62 GP2 ACSM3 LOC81691 UMOD |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |