Assay Details
Target Gene Details
Entrez Gene ID: | 3163 |
Gene Name: | heme oxygenase 2 |
Gene Aliases: |
HO-2 |
Location: |
Chr.16:4474697-4510347 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HMOX2 | NM_001127204.1 | NP_001120676.1 | ||
NM_001127206.2 | NP_001120678.1 | |||
NM_001286267.1 | NP_001273196.1 | |||
NM_001286268.1 | NP_001273197.1 | |||
NM_001286269.1 | NP_001273198.1 | |||
NM_001286271.1 | NP_001273200.1 | |||
NM_002134.3 | NP_002125.3 | |||
XM_017023196.1 | XP_016878685.1 | |||
XM_017023197.1 | XP_016878686.1 | |||
AK222629.1 | BAD96349.1 | |||
AK313660.1 | ||||
BC002396.2 | AAH02396.1 | |||
BE561721.1 | ||||
BI225653.1 | ||||
BM905872.1 | ||||
BP329113.1 | ||||
DA141643.1 | ||||
DA694137.1 | ||||
DB184091.1 | ||||
S34389.1 | AAB22110.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | GLIS2-AS1 RBFOX1 LINC01569 CORO7-PAM16 ADCY9 NAGPA-AS1 NMRAL1 TFAP4 UBN1 DNAJA3 LOC105371063 EEF2KMT GLIS2 GLYR1 SEC14L5 MIR6769A C16orf71 CDIP1 ALG1 PAM16 NUDT16L1 PPL ROGDI MGRN1 SRL C16orf89 VASN SMIM22 C16orf96 NAGPA SEPT12 ANKS3 CORO7 ZNF500 UBALD1 HMOX2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |