Assay Details
Target Gene Details
Entrez Gene ID: | 29904 |
Gene Name: | eukaryotic elongation factor 2 kinase |
Gene Aliases: |
HSU93850, eEF-2K |
Location: |
Chr.16:22206264-22288745 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EEF2K | NM_013302.3 | NP_037434.1 | ||
XM_017023194.1 | XP_016878683.1 | |||
AK308757.1 | ||||
BC032665.1 | AAH32665.1 | |||
DA556486.1 | ||||
DA735877.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469818 | Chr.16:22112125 - 22262126 on Build GRCh38 | Loss | SDR42E2 EEF2K VWA3A |
esv3638195 | Chr.16:22082786 - 22273655 on Build GRCh38 | Gain | SDR42E2 EEF2K VWA3A C16orf52 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |