Assay Details
Target Gene Details
Entrez Gene ID: | 1728 |
Gene Name: | NAD(P)H quinone dehydrogenase 1 |
Gene Aliases: |
DHQU, DIA4, DTD, NMOR1, NMORI, QR1 |
Location: |
Chr.16:69709401-69726668 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NQO1 | NM_000903.2 | NP_000894.1 | ||
NM_001025433.1 | NP_001020604.1 | |||
NM_001025434.1 | NP_001020605.1 | |||
NM_001286137.1 | NP_001273066.1 | |||
AK223050.1 | BAD96770.1 | |||
AK297125.1 | ||||
AK297979.1 | ||||
AK298896.1 | ||||
AK307533.1 | ||||
AK309513.1 | ||||
AK312368.1 | ||||
AK316246.1 | ||||
BC007659.2 | AAH07659.1 | |||
BC107739.1 | AAI07740.1 | |||
BE903613.1 | ||||
CD014005.1 | ||||
DC331440.1 | ||||
J03934.1 | AAA59940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517207 | Chr.16:69711242 - 69738011 on Build GRCh38 | Loss | NQO1 |
esv3892878 | Chr.16:69687512 - 69844258 on Build GRCh38 | Gain | WWP2 NFAT5 NOB1 NQO1 |
esv3638928 | Chr.16:69677077 - 69756070 on Build GRCh38 | Gain | NFAT5 NOB1 NQO1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |