Assay Details
Target Gene Details
Entrez Gene ID: | 101926950 |
Gene Name: | long intergenic non-protein coding RNA 1570 |
Gene Aliases: |
- |
Location: |
Chr.16:5601169-5616250 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC01570 | NR_110902.1 | |||
DB449025.1 |
Target Gene Details
Entrez Gene ID: | 54715 |
Gene Name: | RNA binding protein, fox-1 homolog 1 |
Gene Aliases: |
2BP1, A2BP1, FOX-1, FOX1, HRNBP1 |
Location: |
Chr.16:5239749-7713343 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBFOX1 | XM_017023318.1 | XP_016878807.1 | ||
XM_017023319.1 | XP_016878808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1035156 | Chr.16:5091032 - 5996934 on Build GRCh38 | Gain | RBFOX1 EEF2KMT LINC01570 MIR8065 |
nsv571308 | Chr.16:5535851 - 5969940 on Build GRCh38 | Gain | RBFOX1 LINC01570 MIR8065 |
nsv510675 | Chr.16:5543389 - 5609986 on Build GRCh38 | Deletion | RBFOX1 LINC01570 |
nsv833133 | Chr.16:5533052 - 5681669 on Build GRCh38 | Loss | RBFOX1 LINC01570 MIR8065 |
nsv1048972 | Chr.16:5466061 - 5752374 on Build GRCh38 | Gain | RBFOX1 LINC01570 MIR8065 |
nsv1042301 | Chr.16:5531685 - 5969222 on Build GRCh38 | Gain | RBFOX1 LINC01570 MIR8065 |
esv3892765 | Chr.16:5529214 - 5699442 on Build GRCh38 | Loss | RBFOX1 LINC01570 MIR8065 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |