Assay Details
Target Gene Details
Entrez Gene ID: | 84706 |
Gene Name: | glutamic--pyruvic transaminase 2 |
Gene Aliases: |
ALT2, GPT 2, MRT49 |
Location: |
Chr.16:46884380-46931297 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPT2 | NM_001142466.2 | NP_001135938.1 | ||
NM_133443.3 | NP_597700.1 | |||
AK094971.1 | BAC04465.1 | |||
AY029173.2 | AAK31794.2 | |||
BC062555.1 | AAH62555.1 | |||
DA400677.1 | ||||
HY117879.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3638541 | Chr.16:46727423 - 46897599 on Build GRCh38 | Gain | C16orf87 MYLK3 GPT2 |
nsv1066115 | Chr.16:46429858 - 47284504 on Build GRCh38 | Gain | C16orf87 SHCBP1 DNAJA2 VPS35 NETO2 ANKRD26P1 ITFG1 ITFG1-AS1 MYLK3 GPT2 ORC6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |