Assay Details
Target Gene Details
Entrez Gene ID: | 1800 |
Gene Name: | dipeptidase 1 (renal) |
Gene Aliases: |
MBD1, MDP, RDP |
Location: |
Chr.16:89613308-89641540 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPEP1 | NM_001128141.2 | NP_001121613.1 | ||
NM_004413.3 | NP_004404.1 | |||
XM_005256285.4 | XP_005256342.1 | |||
XM_005256286.3 | XP_005256343.1 | |||
XM_011522925.2 | XP_011521227.1 | |||
XM_017023009.1 | XP_016878498.1 | |||
AK309577.1 | ||||
BM152944.1 | ||||
J05257.1 | AAB59410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | CENPBD1 LOC107984817 GAS8 GAS8-AS1 TUBB3 SPATA33 MC1R TCF25 SPATA2L DEF8 DPEP1 ZNF276 VPS9D1-AS1 VPS9D1 LOC105371419 CHMP1A DBNDD1 SPIRE2 CDK10 AFG3L1P FANCA |
nsv952078 | Chr.16:89602793 - 89640192 on Build GRCh38 | Deletion | DPEP1 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | SPG7 RPL13 LOC101927817 SPATA33 TCF25 ANKRD11 DEF8 CPNE7 DPEP1 ZNF276 VPS9D1 TUBB8P7 CHMP1A DBNDD1 CDK10 AFG3L1P FANCA CENPBD1 FAM157C LOC107984817 URAHP LOC100287036 GAS8 GAS8-AS1 LOC105371414 TUBB3 MC1R SPATA2L VPS9D1-AS1 LOC105371419 SNORD68 SPIRE2 ZNF778 PRDM7 |
nsv573739 | Chr.16:89625890 - 89696855 on Build GRCh38 | Gain | SPATA33 CHMP1A SPATA2L CDK10 DPEP1 |
nsv1063483 | Chr.16:89347250 - 89661187 on Build GRCh38 | Gain | SPG7 RPL13 LOC101927817 SNORD68 SPATA33 CHMP1A ANKRD11 CPNE7 DPEP1 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | SPG7 ACSF3 LOC101927817 SPATA33 SNAI3-AS1 ANKRD11 CPNE7 DPEP1 LOC400558 MVD VPS9D1 PABPN1L CDK10 AFG3L1P FAM157C LOC107984817 URAHP LOC100287036 LOC105371414 GALNS CTU2 CDT1 LOC100289580 VPS9D1-AS1 LINC00304 CDH15 RNF166 LOC105371419 SLC22A31 LOC105371409 ZNF778 PRDM7 RPL13 LOC101927793 TCF25 DEF8 ZNF276 IL17C TUBB8P7 MIR4722 CHMP1A DBNDD1 FANCA CENPBD1 GAS8 GAS8-AS1 LOC100129697 TUBB3 MC1R LOC107987238 SPATA2L TRAPPC2L CBFA2T3 APRT PIEZO1 CYBA SNORD68 SPIRE2 LOC339059 SNAI3 |
nsv509639 | Chr.16:89513538 - 89775915 on Build GRCh38 | Insertion | SPG7 RPL13 SPATA33 SPATA2L CPNE7 DPEP1 ZNF276 VPS9D1-AS1 VPS9D1 SNORD68 CHMP1A CDK10 FANCA |
nsv1066382 | Chr.16:89537178 - 89661187 on Build GRCh38 | Gain | SPG7 RPL13 SNORD68 SPATA33 CHMP1A CPNE7 DPEP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |