Assay Details
Target Gene Details
Entrez Gene ID: | 3163 |
Gene Name: | heme oxygenase 2 |
Gene Aliases: |
HO-2 |
Location: |
Chr.16:4474697-4510347 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HMOX2 | NM_001127204.1 | NP_001120676.1 | ||
NM_001127206.2 | NP_001120678.1 | |||
NM_001286267.1 | NP_001273196.1 | |||
NM_001286268.1 | NP_001273197.1 | |||
NM_001286269.1 | NP_001273198.1 | |||
NM_001286271.1 | NP_001273200.1 | |||
NM_002134.3 | NP_002125.3 | |||
XM_017023196.1 | XP_016878685.1 | |||
XM_017023197.1 | XP_016878686.1 | |||
AK222629.1 | BAD96349.1 | |||
AK313660.1 | ||||
BC002396.2 | AAH02396.1 | |||
BE561721.1 | ||||
BI225653.1 | ||||
BM905872.1 | ||||
BP329113.1 | ||||
DA141643.1 | ||||
DA694137.1 | ||||
DB184091.1 | ||||
S34389.1 | AAB22110.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160332 | Chr.16:4367643 - 4486554 on Build GRCh38 | Duplication | DNAJA3 HMOX2 CORO7-PAM16 VASN CORO7 NMRAL1 |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | NAGPA-AS1 UBALD1 RBFOX1 UBN1 CDIP1 MGRN1 PPL ADCY9 SEC14L5 LINC01569 ALG1 PAM16 HMOX2 C16orf71 GLYR1 SMIM22 TFAP4 MIR6769A CORO7-PAM16 SRL EEF2KMT CORO7 DNAJA3 ZNF500 ROGDI NAGPA LOC105371063 NUDT16L1 GLIS2 VASN C16orf96 C16orf89 SEPT12 ANKS3 GLIS2-AS1 NMRAL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |