Assay Details
Target Gene Details
Entrez Gene ID: | 23322 |
Gene Name: | RPGRIP1 like |
Gene Aliases: |
CORS3, FTM, JBTS7, MKS5, NPHP8, PPP1R134 |
Location: |
Chr.16:53599239-53703934 on Build GRCh38 |
Assay Gene Location: | Within Intron 31 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RPGRIP1L | XM_005255867.1 | XP_005255924.1 | ||
XM_005255868.1 | XP_005255925.1 | |||
XM_017023094.1 | XP_016878583.1 | |||
XM_017023095.1 | XP_016878584.1 | |||
XM_017023096.1 | XP_016878585.1 | |||
XM_017023098.1 | XP_016878587.1 | |||
XM_017023099.1 | XP_016878588.1 | |||
AB023222.1 | BAA76849.1 | |||
AK026552.1 | ||||
AK304790.1 | ||||
AK307929.1 | ||||
BC136433.1 | ||||
CR749645.1 | CAH18439.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |