Assay Details
Target Gene Details
Entrez Gene ID: | 57019 |
Gene Name: | cytokine induced apoptosis inhibitor 1 |
Gene Aliases: |
Anamorsin, DRE2, PRO0915 |
Location: |
Chr.16:57428169-57447528 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CIAPIN1 | NM_001308347.1 | NP_001295276.1 | ||
NM_001308358.1 | NP_001295287.1 | |||
NM_020313.3 | NP_064709.2 | |||
XM_005256063.4 | XP_005256120.1 | |||
AF248964.1 | AAG44562.1 | |||
AK074263.1 | ||||
AK292281.1 | ||||
AL136613.1 | CAB66548.1 | |||
BC002568.2 | AAH02568.1 | |||
BC024196.2 | AAH24196.1 | |||
BC067303.1 | AAH67303.1 | |||
BC071740.1 | AAH71740.1 | |||
BJ991015.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833245 | Chr.16:57434073 - 57584619 on Build GRCh38 | Loss | CIAPIN1 COQ9 ADGRG5 CCDC102A POLR2C DOK4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |