Assay Details
Target Gene Details
Entrez Gene ID: | 123920 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 3 |
Gene Aliases: |
BNAS2, CKLFSF3 |
Location: |
Chr.16:66604032-66613892 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CMTM3 | NM_144601.3 | NP_653202.1 | ||
NM_181553.2 | NP_853531.1 | |||
NR_037613.1 | ||||
NR_037619.1 | ||||
XM_006721131.1 | XP_006721194.1 | |||
XM_011522861.1 | XP_011521163.1 | |||
AF479813.1 | AAN73435.1 | |||
AK056324.1 | BAB71150.1 | |||
AY166714.1 | AAN75573.1 | |||
BC023509.1 | ||||
BC023591.2 | AAH23591.2 | |||
BC036042.1 | AAH36042.2 | |||
BC040227.1 | AAH40227.2 | |||
BC047701.1 | AAH47701.1 | |||
BI457147.1 | ||||
CT001840.1 |
Target Gene Details
Entrez Gene ID: | 146223 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 4 |
Gene Aliases: |
CKLFSF4 |
Location: |
Chr.16:66598178-66696707 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CMTM4 | XM_017022954.1 | XP_016878443.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |