Assay Details
Target Gene Details
Entrez Gene ID: | 6376 |
Gene Name: | C-X3-C motif chemokine ligand 1 |
Gene Aliases: |
ABCD-3, C3Xkine, CXC3, CXC3C, NTN, NTT, SCYD1, fractalkine, neurotactin |
Location: |
Chr.16:57372461-57385048 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CX3CL1 | NM_001304392.1 | 2 | 855 | NP_001291321.1 |
NM_002996.4 | 3 | 976 | NP_002987.1 | |
AB209037.1 | 2 | 3458 | BAD92274.1 | |
AK222692.1 | 3 | 966 | BAD96412.1 | |
AK223351.1 | BAD97071.1 | |||
AK313806.1 | 3 | 947 | ||
BC001163.1 | 3 | 935 | AAH01163.1 | |
BC016164.1 | 3 | 935 | AAH16164.1 | |
CR457426.1 | 3 | 868 | CAG33707.1 | |
U84487.1 | 3 | 947 | AAB49679.1 | |
U91835.1 | 3 | 947 | AAB50014.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |