Assay Details
Target Gene Details
Entrez Gene ID: | 999 |
Gene Name: | cadherin 1 |
Gene Aliases: |
Arc-1, CD324, CDHE, ECAD, LCAM, UVO |
Location: |
Chr.16:68737290-68835542 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH1 | NM_001317184.1 | NP_001304113.1 | ||
NM_001317185.1 | NP_001304114.1 | |||
NM_001317186.1 | NP_001304115.1 | |||
NM_004360.4 | NP_004351.1 | |||
AB025105.1 | BAA88956.1 | |||
AB025106.1 | BAA88957.1 | |||
AK290012.1 | ||||
AK297913.1 | ||||
AK309703.1 | ||||
AK312551.1 | ||||
BC141838.1 | ||||
BC146662.1 | ||||
EU709494.1 | ||||
L08599.1 | AAA61259.1 | |||
Z13009.1 | CAA78353.1 | |||
Z18923.1 | CAA79356.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833267 | Chr.16:68648596 - 68823901 on Build GRCh38 | Gain | CDH1 CDH3 |
nsv457515 | Chr.16:68564104 - 68890598 on Build GRCh38 | Gain | ZFP90 CDH1 CDH3 TANGO6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |