Assay Details
Target Gene Details
Entrez Gene ID: | 55718 |
Gene Name: | RNA polymerase III subunit E |
Gene Aliases: |
RPC5, SIN |
Location: |
Chr.16:22297366-22335103 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLR3E | NM_001258033.1 | NP_001244962.1 | ||
NM_001258034.1 | NP_001244963.1 | |||
NM_001258035.1 | NP_001244964.1 | |||
NM_001258036.1 | NP_001244965.1 | |||
NM_018119.3 | NP_060589.1 | |||
NR_047581.1 | ||||
XM_006721063.2 | XP_006721126.1 | |||
XM_011545892.1 | XP_011544194.1 | |||
XM_011545893.2 | XP_011544195.1 | |||
XM_017023443.1 | XP_016878932.1 | |||
AB040885.1 | BAA95976.1 | |||
AK001371.1 | BAA91655.1 | |||
AK023160.1 | BAB14437.1 | |||
AK023231.1 | BAB14481.1 | |||
AK026645.1 | ||||
AK222671.1 | ||||
AK222789.1 | BAD96509.1 | |||
AK293300.1 | ||||
AK296813.1 | ||||
AK300735.1 | ||||
AY092085.1 | AAM18215.1 | |||
BC000285.1 | AAH00285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3638204 | Chr.16:22296113 - 22352456 on Build GRCh38 | Gain | POLR3E CDR2 |
nsv952951 | Chr.16:22299380 - 22329279 on Build GRCh38 | Deletion | POLR3E |
esv2758637 | Chr.16:22282803 - 22760988 on Build GRCh38 | Gain+Loss | EEF2K POLR3E LOC653786 LOC440346 NPIPB5 RRN3P3 CDR2 SMG1P1 MFSD13B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |