Assay Details
Target Gene Details
Entrez Gene ID: | 23295 |
Gene Name: | mahogunin ring finger 1 |
Gene Aliases: |
RNF156 |
Location: |
Chr.16:4616493-4690974 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MGRN1 | NM_001142289.2 | NP_001135761.2 | ||
NM_001142290.2 | NP_001135762.1 | |||
NM_001142291.2 | NP_001135763.2 | |||
NM_015246.3 | NP_056061.1 | |||
NR_102267.1 | ||||
XM_005255217.1 | XP_005255274.1 | |||
XM_005255218.1 | XP_005255275.1 | |||
XM_005255219.3 | XP_005255276.1 | |||
XM_005255220.3 | XP_005255277.1 | |||
XM_005255221.3 | XP_005255278.1 | |||
XM_011522441.1 | XP_011520743.1 | |||
XM_017023091.1 | XP_016878580.1 | |||
XM_017023092.1 | XP_016878581.1 | |||
XM_017023093.1 | XP_016878582.1 | |||
AB011116.1 | ||||
AK299052.1 | ||||
BC050389.1 | AAH50389.1 | |||
CN314787.1 | ||||
DA118421.1 | ||||
DA590130.1 | ||||
EF471397.2 | ||||
EF471398.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160333 | Chr.16:4570296 - 4709737 on Build GRCh38 | Deletion | C16orf96 MIR6769A NUDT16L1 MGRN1 UBALD1 ANKS3 |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | PPL RBFOX1 DNAJA3 EEF2KMT ROGDI UBALD1 LOC105371063 ADCY9 TFAP4 GLIS2 CORO7 NMRAL1 GLYR1 MGRN1 C16orf89 ZNF500 ANKS3 LINC01569 GLIS2-AS1 NAGPA-AS1 PAM16 UBN1 MIR6769A C16orf71 SRL HMOX2 ALG1 SEC14L5 CORO7-PAM16 SEPT12 C16orf96 VASN NUDT16L1 SMIM22 CDIP1 NAGPA |
nsv1050237 | Chr.16:4641028 - 4744872 on Build GRCh38 | Loss | MIR6769A NUDT16L1 C16orf71 MGRN1 ANKS3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |