Assay Details
Target Gene Details
Entrez Gene ID: | 2775 |
Gene Name: | G protein subunit alpha o1 |
Gene Aliases: |
EIEE17, G-ALPHA-o, GNAO, HLA-DQB1 |
Location: |
Chr.16:56191339-56357444 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNAO1 | NM_020988.2 | NP_066268.1 | ||
NM_138736.2 | NP_620073.2 | |||
XM_011523003.2 | XP_011521305.1 | |||
AF493894.1 | AAM12608.1 | |||
AF493895.1 | AAM12609.1 | |||
AK056008.1 | ||||
BC030027.2 | AAH30027.2 | |||
CR936770.1 | ||||
KR081949.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |