Assay Details
Target Gene Details
Entrez Gene ID: | 3931 |
Gene Name: | lecithin-cholesterol acyltransferase |
Gene Aliases: |
- |
Location: |
Chr.16:67939884-67944112 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LCAT | NM_000229.1 | NP_000220.1 | ||
BC014781.1 | AAH14781.1 | |||
BT009748.1 | AAP88750.1 | |||
M12625.1 | AAA59498.1 | |||
M26268.1 | AAA59499.1 | |||
X06537.1 | CAB56610.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | PSMB10 RANBP10 EXOC3L1 CENPT PLEKHG4 NRN1L PSKH1 NUTF2 PARD6A ACD LOC107984813 LOC100131303 DPEP3 FAM65A TSNAXIP1 TPPP3 AGRP TRADD CARMIL2 LRRC29 EDC4 LCAT ENKD1 LRRC36 SLC12A4 THAP11 C16orf86 DPEP2 CTCF ATP6V0D1 KIAA0895L FHOD1 TMEM208 LOC100505942 NOL3 FBXL8 HSD11B2 E2F4 LOC105369155 ZDHHC1 HSF4 CTRL KCTD19 MIR328 SLC9A5 DUS2 GFOD2 ELMO3 DDX28 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | B3GNT9 PSMB10 RANBP10 EXOC3L1 CENPT PLEKHG4 NRN1L PSKH1 NUTF2 PARD6A ACD LOC107984813 LOC100131303 DPEP3 FAM65A TSNAXIP1 TPPP3 AGRP TRADD CARMIL2 LRRC29 EDC4 LCAT ENKD1 LRRC36 SLC12A4 THAP11 C16orf86 DPEP2 CTCF ATP6V0D1 KIAA0895L C16orf70 FHOD1 TMEM208 LOC100505942 NOL3 FBXL8 HSD11B2 E2F4 LOC105369155 ZDHHC1 HSF4 CTRL KCTD19 MIR328 SLC9A5 DUS2 GFOD2 ELMO3 DDX28 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |