Assay Details
Target Gene Details
Entrez Gene ID: | 1800 |
Gene Name: | dipeptidase 1 (renal) |
Gene Aliases: |
MBD1, MDP, RDP |
Location: |
Chr.16:89613308-89641540 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPEP1 | NM_001128141.2 | NP_001121613.1 | ||
NM_004413.3 | NP_004404.1 | |||
XM_005256285.4 | XP_005256342.1 | |||
XM_005256286.3 | XP_005256343.1 | |||
XM_011522925.2 | XP_011521227.1 | |||
XM_017023009.1 | XP_016878498.1 | |||
AK309577.1 | ||||
BC017023.1 | AAH17023.1 | |||
BM152944.1 | ||||
BT006664.1 | AAP35310.1 | |||
CR457404.1 | CAG33685.1 | |||
D13138.1 | BAA02431.1 | |||
J05257.1 | AAB59410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | SPATA33 TUBB3 GAS8-AS1 SPIRE2 CENPBD1 MC1R VPS9D1-AS1 DEF8 GAS8 CDK10 SPATA2L LOC105371419 DPEP1 VPS9D1 FANCA ZNF276 DBNDD1 CHMP1A TCF25 LOC107984817 AFG3L1P |
nsv952078 | Chr.16:89602793 - 89640192 on Build GRCh38 | Deletion | DPEP1 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | SPATA33 GAS8-AS1 MC1R DEF8 LOC100287036 CPNE7 VPS9D1 FANCA ZNF276 ANKRD11 LOC107984817 LOC101927817 RPL13 AFG3L1P TUBB3 SPIRE2 SPG7 FAM157C CENPBD1 VPS9D1-AS1 TUBB8P7 GAS8 CDK10 SPATA2L LOC105371419 SNORD68 URAHP ZNF778 DPEP1 LOC105371414 PRDM7 DBNDD1 CHMP1A TCF25 |
nsv573739 | Chr.16:89625890 - 89696855 on Build GRCh38 | Gain | DPEP1 SPATA33 CHMP1A CDK10 SPATA2L |
nsv1063483 | Chr.16:89347250 - 89661187 on Build GRCh38 | Gain | DPEP1 SPATA33 CPNE7 SPG7 ANKRD11 CHMP1A LOC101927817 RPL13 SNORD68 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | SPATA33 LOC339059 CBFA2T3 GAS8-AS1 SNAI3-AS1 SLC22A31 CTU2 MVD ACSF3 LOC100287036 RNF166 CPNE7 VPS9D1 LINC00304 ANKRD11 LOC107984817 LOC101927817 RPL13 PIEZO1 AFG3L1P SPIRE2 SPG7 LOC100289580 CENPBD1 VPS9D1-AS1 TUBB8P7 IL17C GAS8 CDK10 LOC400558 DPEP1 LOC105371414 CHMP1A TCF25 TRAPPC2L GALNS MC1R LOC107987238 DEF8 SNAI3 LOC105371409 MIR4722 FANCA ZNF276 APRT TUBB3 FAM157C PABPN1L SPATA2L LOC105371419 SNORD68 CYBA URAHP ZNF778 LOC100129697 CDT1 PRDM7 DBNDD1 CDH15 LOC101927793 |
nsv509639 | Chr.16:89513538 - 89775915 on Build GRCh38 | Insertion | SPATA33 SPG7 VPS9D1-AS1 CDK10 SPATA2L SNORD68 DPEP1 CPNE7 VPS9D1 FANCA ZNF276 CHMP1A RPL13 |
nsv1066382 | Chr.16:89537178 - 89661187 on Build GRCh38 | Gain | DPEP1 SPATA33 CPNE7 SPG7 CHMP1A RPL13 SNORD68 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |