Assay Details
Target Gene Details
Entrez Gene ID: | 81533 |
Gene Name: | integrin alpha FG-GAP repeat containing 1 |
Gene Aliases: |
2310047C21Rik, CDA08, LNKN-1, TIP |
Location: |
Chr.16:47154387-47461274 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ITFG1 | NM_001305002.1 | NP_001291931.1 | ||
NM_030790.4 | NP_110417.2 | |||
AF212247.1 | AAG41782.1 | |||
AF503339.1 | AAN31655.1 | |||
AK027596.1 | BAB55220.1 | |||
AK295228.1 | ||||
AK299641.1 | ||||
AK301908.1 | ||||
AL833705.1 | ||||
BC006321.2 | AAH06321.2 | |||
BC024162.1 | AAH24162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525170 | Chr.16:47160140 - 47269724 on Build GRCh38 | Gain | ITFG1 ITFG1-AS1 |
nsv528423 | Chr.16:47129963 - 47313869 on Build GRCh38 | Loss | ITFG1 ITFG1-AS1 NETO2 |
nsv1066115 | Chr.16:46429858 - 47284504 on Build GRCh38 | Gain | ANKRD26P1 GPT2 ITFG1 MYLK3 ITFG1-AS1 DNAJA2 NETO2 SHCBP1 C16orf87 VPS35 ORC6 |