Assay Details
Target Gene Details
Entrez Gene ID: | 51704 |
Gene Name: | G protein-coupled receptor class C group 5 member B |
Gene Aliases: |
RAIG-2, RAIG2 |
Location: |
Chr.16:19858971-19886045 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPRC5B | NM_001304771.1 | NP_001291700.1 | ||
NM_016235.2 | NP_057319.1 | |||
XM_006721051.2 | XP_006721114.1 | |||
XM_006721052.2 | XP_006721115.1 | |||
AF181862.1 | AAF05331.1 | |||
AF202640.1 | AAF67321.1 | |||
AJ276101.1 | CAC00632.1 | |||
AK075119.1 | BAC11414.1 | |||
AK294680.1 | ||||
AK295204.1 | ||||
AK296754.1 | ||||
AK302838.1 | ||||
AK315989.1 | ||||
BC034467.2 | AAH34467.1 | |||
FJ529380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428321 | Chr.16:19742033 - 19876673 on Build GRCh38 | Loss |
![]() ![]() |
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)