Assay Details
Target Gene Details
Entrez Gene ID: | 1800 |
Gene Name: | dipeptidase 1 (renal) |
Gene Aliases: |
MBD1, MDP, RDP |
Location: |
Chr.16:89613308-89641540 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPEP1 | NM_001128141.2 | NP_001121613.1 | ||
NM_004413.3 | NP_004404.1 | |||
XM_005256285.4 | XP_005256342.1 | |||
XM_005256286.3 | XP_005256343.1 | |||
XM_011522925.2 | XP_011521227.1 | |||
XM_017023009.1 | XP_016878498.1 | |||
AK309577.1 | ||||
BC017023.1 | AAH17023.1 | |||
BM152944.1 | ||||
BT006664.1 | AAP35310.1 | |||
CR457404.1 | CAG33685.1 | |||
D13138.1 | BAA02431.1 | |||
J05257.1 | AAB59410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | FANCA CHMP1A GAS8-AS1 DBNDD1 TCF25 MC1R ZNF276 VPS9D1 SPATA2L CENPBD1 GAS8 LOC105371419 CDK10 VPS9D1-AS1 DEF8 SPIRE2 DPEP1 AFG3L1P LOC107984817 SPATA33 TUBB3 |
nsv952078 | Chr.16:89602793 - 89640192 on Build GRCh38 | Deletion | DPEP1 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | URAHP ZNF778 DBNDD1 ANKRD11 VPS9D1 LOC105371414 CENPBD1 CDK10 FAM157C VPS9D1-AS1 TUBB8P7 LOC101927817 SNORD68 LOC107984817 SPATA33 CPNE7 LOC100287036 FANCA SPG7 CHMP1A GAS8-AS1 TCF25 MC1R ZNF276 SPATA2L GAS8 LOC105371419 DEF8 SPIRE2 DPEP1 AFG3L1P TUBB3 RPL13 PRDM7 |
nsv573739 | Chr.16:89625890 - 89696855 on Build GRCh38 | Gain | DPEP1 CHMP1A SPATA33 SPATA2L CDK10 |
nsv1063483 | Chr.16:89347250 - 89661187 on Build GRCh38 | Gain | DPEP1 SPG7 CHMP1A LOC101927817 SNORD68 ANKRD11 SPATA33 CPNE7 RPL13 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | LOC105371409 ANKRD11 VPS9D1 CENPBD1 VPS9D1-AS1 LOC101927817 LOC107984817 LOC100289580 CPNE7 LOC101927793 SNAI3 SPG7 TCF25 RNF166 ZNF276 ACSF3 SPATA2L LOC105371419 DEF8 PABPN1L APRT IL17C RPL13 LOC100129697 PRDM7 URAHP ZNF778 LINC00304 DBNDD1 CYBA LOC105371414 CDK10 FAM157C TUBB8P7 SNORD68 LOC400558 CDT1 CDH15 SNAI3-AS1 MIR4722 SPATA33 LOC339059 TRAPPC2L GALNS PIEZO1 LOC100287036 MVD FANCA CHMP1A GAS8-AS1 MC1R GAS8 CBFA2T3 SPIRE2 DPEP1 SLC22A31 AFG3L1P CTU2 TUBB3 LOC107987238 |
nsv509639 | Chr.16:89513538 - 89775915 on Build GRCh38 | Insertion | FANCA SPG7 CHMP1A ZNF276 VPS9D1 SPATA2L CDK10 VPS9D1-AS1 DPEP1 SNORD68 SPATA33 CPNE7 RPL13 |
nsv1066382 | Chr.16:89537178 - 89661187 on Build GRCh38 | Gain | DPEP1 SPG7 CHMP1A SNORD68 SPATA33 CPNE7 RPL13 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |