Assay Details
Target Gene Details
Entrez Gene ID: | 3163 |
Gene Name: | heme oxygenase 2 |
Gene Aliases: |
HO-2 |
Location: |
Chr.16:4474697-4510347 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HMOX2 | NM_001127204.1 | NP_001120676.1 | ||
NM_001127206.2 | NP_001120678.1 | |||
NM_001286267.1 | NP_001273196.1 | |||
NM_001286268.1 | NP_001273197.1 | |||
NM_001286269.1 | NP_001273198.1 | |||
NM_001286271.1 | NP_001273200.1 | |||
NM_002134.3 | NP_002125.3 | |||
XM_017023196.1 | XP_016878685.1 | |||
XM_017023197.1 | XP_016878686.1 | |||
AK222629.1 | BAD96349.1 | |||
AK313660.1 | ||||
BC002396.2 | AAH02396.1 | |||
BE561721.1 | ||||
BI225653.1 | ||||
BM905872.1 | ||||
BP329113.1 | ||||
DA141643.1 | ||||
DA694137.1 | ||||
DB184091.1 | ||||
S34389.1 | AAB22110.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | EEF2KMT NAGPA-AS1 ROGDI HMOX2 UBALD1 SEPT12 GLIS2-AS1 SEC14L5 UBN1 ANKS3 MGRN1 NAGPA CORO7-PAM16 TFAP4 CORO7 CDIP1 C16orf89 NMRAL1 PAM16 VASN MIR6769A SRL NUDT16L1 ALG1 C16orf71 ZNF500 LINC01569 GLYR1 SMIM22 DNAJA3 GLIS2 RBFOX1 ADCY9 C16orf96 PPL LOC105371063 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |