Assay Details
Target Gene Details
Entrez Gene ID: | 81831 |
Gene Name: | neuropilin and tolloid like 2 |
Gene Aliases: |
BTCL2, NEOT2 |
Location: |
Chr.16:47081520-47144025 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NETO2 | NM_001201477.1 | NP_001188406.1 | ||
NM_018092.4 | NP_060562.3 | |||
XM_006721289.1 | XP_006721352.1 | |||
XM_006721291.1 | XP_006721354.1 | |||
XM_006721292.1 | XP_006721355.1 | |||
XM_017023740.1 | XP_016879229.1 | |||
AK074937.1 | BAC11303.1 | |||
AK297408.1 | ||||
AL834404.1 | CAD39066.1 | |||
AY358718.1 | AAQ89080.1 | |||
BC108727.1 | AAI08728.1 | |||
BX538063.1 | CAD97994.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066115 | Chr.16:46429858 - 47284504 on Build GRCh38 | Gain | SHCBP1 VPS35 MYLK3 ITFG1-AS1 ORC6 GPT2 ITFG1 DNAJA2 ANKRD26P1 C16orf87 NETO2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |