Assay Details
Target Gene Details
Entrez Gene ID: | 9057 |
Gene Name: | solute carrier family 7 member 6 |
Gene Aliases: |
LAT-2, LAT3, y+LAT-2 |
Location: |
Chr.16:68263830-68301823 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC7A6 | NM_001076785.2 | NP_001070253.1 | ||
NM_003983.5 | NP_003974.3 | |||
XM_011523433.1 | XP_011521735.1 | |||
XM_011523434.1 | XP_011521736.1 | |||
XM_011523438.1 | XP_011521740.1 | |||
XM_011523439.1 | XP_011521741.1 | |||
XM_017023851.1 | XP_016879340.1 | |||
AK310866.1 | ||||
AK311610.1 | ||||
BC028216.1 | AAH28216.1 | |||
CR749291.1 | CAH18146.1 | |||
CR749475.1 | CAH18305.1 | |||
D87432.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv510691 | Chr.16:68266527 - 68325431 on Build GRCh38 | Deletion | PRMT7 SLC7A6OS SLC7A6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |