Assay Details
Target Gene Details
Entrez Gene ID: | 94160 |
Gene Name: | ATP binding cassette subfamily C member 12 |
Gene Aliases: |
MRP9 |
Location: |
Chr.16:48082973-48146770 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABCC12 | NM_033226.2 | NP_150229.2 | ||
AF395908.1 | AAL79528.1 | |||
AF395909.1 | AAL79529.1 | |||
AF411577.1 | AAL99900.1 | |||
AF411578.1 | AAL99901.1 | |||
AK307109.1 | ||||
AK308599.1 | ||||
AY040220.1 | AAK76740.1 | |||
AY196326.1 | AAO40749.1 | |||
DA096073.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2761906 | Chr.16:48075816 - 48091944 on Build GRCh38 | Loss | ABCC12 |
nsv520355 | Chr.16:47390917 - 48130866 on Build GRCh38 | Gain | LOC100507534 ITFG1 LOC101927132 PHKB ABCC12 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |