Assay Details
Target Gene Details
Entrez Gene ID: | 1800 |
Gene Name: | dipeptidase 1 (renal) |
Gene Aliases: |
MBD1, MDP, RDP |
Location: |
Chr.16:89613308-89641540 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPEP1 | NM_001128141.2 | NP_001121613.1 | ||
NM_004413.3 | NP_004404.1 | |||
XM_005256285.4 | XP_005256342.1 | |||
XM_005256286.3 | XP_005256343.1 | |||
XM_011522925.2 | XP_011521227.1 | |||
XM_017023009.1 | XP_016878498.1 | |||
AK309577.1 | ||||
BM152944.1 | ||||
J05257.1 | AAB59410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | LOC107984817 DEF8 SPIRE2 CHMP1A AFG3L1P MC1R ZNF276 GAS8-AS1 VPS9D1-AS1 DPEP1 FANCA DBNDD1 SPATA2L CDK10 SPATA33 CENPBD1 TCF25 LOC105371419 GAS8 VPS9D1 TUBB3 |
nsv952078 | Chr.16:89602793 - 89640192 on Build GRCh38 | Deletion | DPEP1 |
nsv1929 | Chr.16:89601681 - 89628517 on Build GRCh38 | Insertion | DPEP1 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | LOC107984817 DEF8 CHMP1A ZNF276 FAM157C FANCA DBNDD1 RPL13 CDK10 SPATA33 CENPBD1 SPG7 LOC105371419 LOC101927817 TUBB8P7 TUBB3 SNORD68 SPIRE2 AFG3L1P MC1R GAS8-AS1 URAHP VPS9D1-AS1 ANKRD11 DPEP1 CPNE7 SPATA2L TCF25 ZNF778 GAS8 LOC100287036 VPS9D1 LOC105371414 PRDM7 |
nsv573738 | Chr.16:89549357 - 89625890 on Build GRCh38 | Loss | CPNE7 SNORD68 RPL13 SPG7 DPEP1 |
nsv1063483 | Chr.16:89347250 - 89661187 on Build GRCh38 | Gain | CPNE7 SNORD68 RPL13 CHMP1A SPATA33 SPG7 ANKRD11 DPEP1 LOC101927817 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | LOC107984817 SNAI3 LINC00304 CHMP1A ZNF276 CYBA CDT1 RPL13 CENPBD1 SPG7 LOC105371419 SNAI3-AS1 TUBB8P7 ACSF3 LOC100289580 SPIRE2 PABPN1L AFG3L1P CTU2 MC1R SLC22A31 GAS8-AS1 URAHP ANKRD11 IL17C LOC107987238 CBFA2T3 TRAPPC2L LOC100129697 LOC100287036 APRT VPS9D1 PIEZO1 LOC105371414 PRDM7 DEF8 GALNS LOC101927793 MIR4722 LOC339059 FAM157C FANCA DBNDD1 CDK10 SPATA33 LOC400558 LOC105371409 LOC101927817 TUBB3 RNF166 SNORD68 VPS9D1-AS1 DPEP1 CPNE7 CDH15 SPATA2L MVD TCF25 ZNF778 GAS8 |
nsv509639 | Chr.16:89513538 - 89775915 on Build GRCh38 | Insertion | SNORD68 CHMP1A ZNF276 VPS9D1-AS1 DPEP1 CPNE7 FANCA SPATA2L RPL13 CDK10 SPATA33 SPG7 VPS9D1 |
nsv1066382 | Chr.16:89537178 - 89661187 on Build GRCh38 | Gain | CPNE7 SNORD68 RPL13 CHMP1A SPATA33 SPG7 DPEP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |