Assay Details
Target Gene Details
Entrez Gene ID: | 6299 |
Gene Name: | spalt like transcription factor 1 |
Gene Aliases: |
HEL-S-89, HSAL1, Sal-1, TBS, ZNF794 |
Location: |
Chr.16:51135975-51152386 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SALL1 | NM_001127892.1 | NP_001121364.1 | ||
NM_002968.2 | NP_002959.2 | |||
XM_006721241.3 | XP_006721304.1 | |||
XM_011523254.2 | XP_011521556.1 | |||
AK295103.1 | ||||
AK307835.1 | ||||
BC113881.1 | ||||
BC113906.1 | ||||
BC131548.1 | ||||
BP229682.1 | ||||
DA318592.1 | ||||
EU668343.1 | ||||
Y18265.1 | CAB41400.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833224 | Chr.16:51062577 - 51219186 on Build GRCh38 | Loss | SALL1 LOC105371253 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |