Assay Details
Target Gene Details
Entrez Gene ID: | 100529144 |
Gene Name: | CORO7-PAM16 readthrough |
Gene Aliases: |
- |
Location: |
Chr.16:4340251-4416961 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CORO7-PAM16 | NM_001201479.1 | NP_001188408.1 | ||
AL833954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471075 | Chr.16:4317729 - 4395326 on Build GRCh38 | Loss | GLIS2-AS1 CORO7 CORO7-PAM16 GLIS2 VASN PAM16 |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | UBN1 LOC105371063 LINC01569 ANKS3 ZNF500 NAGPA-AS1 RBFOX1 ALG1 UBALD1 SEC14L5 C16orf71 DNAJA3 PPL EEF2KMT CORO7-PAM16 NAGPA SEPT12 GLIS2-AS1 NUDT16L1 ADCY9 CDIP1 NMRAL1 C16orf96 GLIS2 VASN PAM16 SRL ROGDI GLYR1 CORO7 SMIM22 HMOX2 TFAP4 MIR6769A MGRN1 C16orf89 |
nsv571274 | Chr.16:4309420 - 4441189 on Build GRCh38 | Gain | GLIS2-AS1 CORO7 DNAJA3 CORO7-PAM16 GLIS2 VASN PAM16 |
nsv527866 | Chr.16:4206462 - 4412618 on Build GRCh38 | Loss | GLIS2-AS1 LOC105371063 CORO7 LINC01569 TFAP4 CORO7-PAM16 GLIS2 VASN PAM16 SRL |
esv3584924 | Chr.16:4330628 - 4469258 on Build GRCh38 | Gain | CORO7 DNAJA3 CORO7-PAM16 NMRAL1 GLIS2 VASN PAM16 |
esv23732 | Chr.16:4340813 - 4476244 on Build GRCh38 | Gain | CORO7 HMOX2 DNAJA3 CORO7-PAM16 NMRAL1 VASN PAM16 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |