Assay Details
Target Gene Details
Entrez Gene ID: | 60490 |
Gene Name: | phosphopantothenoylcysteine decarboxylase |
Gene Aliases: |
MDS018, PPC-DC, coaC |
Location: |
Chr.15:75023533-75060180 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPCDC | NM_001301101.1 | NP_001288030.1 | ||
NM_001301102.1 | NP_001288031.1 | |||
NM_001301103.1 | NP_001288032.1 | |||
NM_021823.4 | NP_068595.3 | |||
NR_125369.1 | ||||
XM_017022461.1 | XP_016877950.1 | |||
XM_017022464.1 | XP_016877953.1 | |||
XM_017022465.1 | XP_016877954.1 | |||
AK027491.1 | BAB55151.1 | |||
AY358848.1 | AAQ89207.1 | |||
BC014409.1 | AAH14409.1 | |||
BF243417.1 | ||||
BI833814.1 | ||||
BU500301.1 | ||||
DB488540.1 | ||||
DW437029.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569993 | Chr.15:74949814 - 75191411 on Build GRCh38 | Gain | SCAMP5 PPCDC RPP25 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | MIR6882 C15orf39 NPTN CYP11A1 LOC105376731 PML REC114 LMAN1L LOC107984728 TBC1D21 MIR6881 SEMA7A CPLX3 GOLGA6C HIGD2B CCDC33 LOC101929333 LOXL1 C15orf59 UBL7 STOML1 SCAMP2 CYP1A2 RPP25 EDC3 CD276 HCN4 BBS4 ULK3 COX5A PPCDC ARID3B FAM219B NEO1 ISLR CSK LOC732265 LOC283731 ADPGK-AS1 SCAMP5 ADPGK LOC102723750 C15orf59-AS1 LOXL1-AS1 MIR4513 STRA6 CLK3 ISLR2 MPI CYP1A1 LOC729739 NPTN-IT1 GOLGA6A UBL7-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |