Assay Details
Target Gene Details
Entrez Gene ID: | 4756 |
Gene Name: | neogenin 1 |
Gene Aliases: |
IGDCC2, NGN, NTN1R2 |
Location: |
Chr.15:73051715-73305206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525462 | Chr.15:73057292 - 73271154 on Build GRCh38 | Loss | NEO1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | ISLR2 LOC102723750 TBC1D21 C15orf59-AS1 GOLGA6C NPTN LOC107984728 BBS4 GOLGA6A HCN4 C15orf59 RPP25 HIGD2B LOC729739 COX5A UBL7 C15orf39 ARID3B CYP1A1 LOC283731 SEMA7A ULK3 ISLR MIR4513 CCDC33 NPTN-IT1 NEO1 CPLX3 MIR6882 REC114 LOC105376731 LOC732265 STRA6 MPI STOML1 LOC101929333 CYP1A2 EDC3 ADPGK CD276 PML SCAMP5 CYP11A1 CLK3 PPCDC CSK ADPGK-AS1 LMAN1L SCAMP2 MIR6881 UBL7-AS1 LOXL1 FAM219B LOXL1-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |