Assay Details
Target Gene Details
Entrez Gene ID: | 60490 |
Gene Name: | phosphopantothenoylcysteine decarboxylase |
Gene Aliases: |
MDS018, PPC-DC, coaC |
Location: |
Chr.15:75023533-75060180 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPCDC | NM_001301101.1 | NP_001288030.1 | ||
NM_001301102.1 | NP_001288031.1 | |||
NM_001301103.1 | NP_001288032.1 | |||
NM_021823.4 | NP_068595.3 | |||
NR_125369.1 | ||||
XM_017022461.1 | XP_016877950.1 | |||
XM_017022464.1 | XP_016877953.1 | |||
XM_017022465.1 | XP_016877954.1 | |||
AK027491.1 | BAB55151.1 | |||
AY358848.1 | AAQ89207.1 | |||
BC014409.1 | AAH14409.1 | |||
BF243417.1 | ||||
BI833814.1 | ||||
BU500301.1 | ||||
DB488540.1 | ||||
DW437029.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv569993 | Chr.15:74949814 - 75191411 on Build GRCh38 | Gain | PPCDC RPP25 SCAMP5 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | SEMA7A CYP1A1 LOXL1 ADPGK PML CSK LOC101929333 LOC102723750 MPI UBL7-AS1 UBL7 LOC283731 NEO1 PPCDC LOXL1-AS1 CCDC33 RPP25 ADPGK-AS1 LMAN1L CLK3 LOC729739 LOC732265 EDC3 HIGD2B LOC105376731 ARID3B ULK3 CPLX3 BBS4 NPTN-IT1 ISLR2 STRA6 SCAMP2 LOC107984728 CYP1A2 C15orf59-AS1 CYP11A1 CD276 TBC1D21 MIR6882 GOLGA6C C15orf39 FAM219B SCAMP5 STOML1 C15orf59 MIR4513 COX5A HCN4 REC114 NPTN GOLGA6A MIR6881 ISLR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |