Assay Details
Target Gene Details
Entrez Gene ID: | 4756 |
Gene Name: | neogenin 1 |
Gene Aliases: |
IGDCC2, NGN, NTN1R2 |
Location: |
Chr.15:73051715-73305206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525462 | Chr.15:73057292 - 73271154 on Build GRCh38 | Loss | NEO1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | CSK STOML1 MIR4513 NPTN LOC107984728 COX5A BBS4 GOLGA6A REC114 CYP11A1 CYP1A2 ISLR2 FAM219B CLK3 MIR6881 SEMA7A LOC102723750 LOC101929333 HIGD2B C15orf59 HCN4 CCDC33 MIR6882 RPP25 C15orf59-AS1 LOC729739 ISLR ULK3 LOC732265 LOC105376731 LOC283731 GOLGA6C EDC3 C15orf39 STRA6 LOXL1 SCAMP5 ADPGK UBL7 ARID3B LMAN1L NEO1 LOXL1-AS1 CD276 MPI CYP1A1 CPLX3 UBL7-AS1 PML PPCDC TBC1D21 ADPGK-AS1 SCAMP2 NPTN-IT1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |