Assay Details
Target Gene Details
Entrez Gene ID: | 6692 |
Gene Name: | serine peptidase inhibitor, Kunitz type 1 |
Gene Aliases: |
HAI, HAI1, MANSC2 |
Location: |
Chr.15:40844018-40857655 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPINT1 | NM_001032367.1 | NP_001027539.1 | ||
NM_003710.3 | NP_003701.1 | |||
NM_181642.2 | NP_857593.1 | |||
XM_006720657.1 | XP_006720720.1 | |||
XM_011521957.1 | XP_011520259.1 | |||
XM_011521958.1 | XP_011520260.1 | |||
AB000095.1 | BAA25014.1 | |||
AK300195.1 | ||||
AK303899.1 | ||||
AK314825.1 | ||||
AY296715.1 | AAP44001.1 | |||
AY358969.1 | AAQ89328.1 | |||
BC004140.1 | AAH04140.1 | |||
BC018702.1 | AAH18702.1 | |||
BT007425.1 | AAP36093.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1054764 | Chr.15:40776427 - 40904212 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv569228 | Chr.15:40814287 - 40901953 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)