Assay Details
Target Gene Details
Entrez Gene ID: | 58472 |
Gene Name: | sulfide quinone reductase-like (yeast) |
Gene Aliases: |
CGI-44, PRO1975, SQOR |
Location: |
Chr.15:45631148-45691294 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SQRDL | NM_001271213.1 | NP_001258142.1 | ||
NM_021199.3 | NP_067022.1 | |||
AF042284.1 | AAD41160.1 | |||
AF151802.1 | AAD34039.1 | |||
AK130140.1 | ||||
AK222463.1 | BAD96183.1 | |||
AK313111.1 | ||||
BC016836.1 | AAH16836.1 | |||
DA821218.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892684 | Chr.15:45307172 - 46327637 on Build GRCh38 | Gain | SQRDL HMGN2P46 BLOC1S6 C15orf48 GATM LOC105370802 MIR147B LOC105376714 SLC30A4 SPATA5L1 |
esv3569553 | Chr.15:45366990 - 46080626 on Build GRCh38 | Gain | SQRDL HMGN2P46 BLOC1S6 C15orf48 GATM LOC105370802 MIR147B LOC105376714 SLC30A4 SPATA5L1 |
nsv1160292 | Chr.15:45351161 - 46175480 on Build GRCh38 | Duplication | SQRDL HMGN2P46 BLOC1S6 C15orf48 GATM LOC105370802 MIR147B LOC105376714 SLC30A4 SPATA5L1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |